missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoMag 96 Blood

Designed for the rapid parallel purification of genomic DNA from whole blood in a 96-well format.

Brand:  Macherey-Nagel Bioanalysis™ 744500.4


 View more versions of this product

Product Code. 12758672

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

After lysis of the blood cells genomic DNA is selectively bound to the NucleoMag Blood beads. Contaminants are washed away and the DNA can be eluted from the beads under low-salt conditions. As the magnetic bead technology provides a closed system (no transfer of sample from one well to another is necessary during the preparation) the risk for cross-contamination is greatly reduced. The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg. NucleoMag SEP) or magnetic separators integrated into robotic workstations.Kit components: NucleoMag B-Beads, elution plate (u-bottom), buffers.

  • Magnetic bead technology
  • Yield: 1 to 4 μg
  • Sample material: 100 μL whole blood
  • Elution volume: 25 to 100 μL
  • Binding capacity: 0.4 μL/ 1 μL beads
  • Preparation time: 120 min/96 preps
  • Fragment size: 300 bp to 30 kb
  • Format: superparamagnetic beads (size of beads: 1 to 3 μm)
Specifications

Specifications

384 Preps
Highly Reactive Superparamagnetic Beads (1 to 3 μm)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Category
  • Flammable liquid Category 3

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.