Learn More
Macherey-Nagel Bioanalysis™ NucleoSEQ™ Kit
NucleoSEQ™ Kit for dye terminator removal.
Brand:  Macherey-Nagel Bioanalysis™ 740523.250
 View more versions of this product
Product Code. 10373438
  • 476.00€ / 250 reactions
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Contains NucleoSEQ™ columns, collection tubes, and a user manual
  • Designed for fast and effective clean-up of nucleic acids
  • Using gel exclusion in a convenient spin column format allows reliable removal of smaller molecules from nucleic acids
  • Impurities, for example, salts, excess of labels, nucleotides, traces of organic solvents, primers are retained by the column while nucleic acids of interest are recovered with high yield
  • The columns are pre-filled with size exclusion matrix
  • Maximum sample volume to be loaded onto the column: 20 μL
  • Columns with dry gel matrix can be stored at room temperature for 12 months
  • Columns with hydrated gel matrix can be stored at 4°C for 14 days
Specifications
Specifications
250 Preps
5 min/prep (without hydration of matrix)
Dye terminator removal