Learn More
Macherey-Nagel Bioanalysis™ NucleoSEQ™ Kit
NucleoSEQ™ Kit for dye terminator removal.
Brand: Macherey-Nagel Bioanalysis™ 740523.10
This item is not returnable.
View return policy
Description
- Contains NucleoSEQ™ columns, collection tubes, and a user manual
- Designed for fast and effective clean-up of nucleic acids
- Using gel exclusion in a convenient spin column format allows reliable removal of smaller molecules from nucleic acids
- Impurities, for example, salts, excess of labels, nucleotides, traces of organic solvents, primers are retained by the column while nucleic acids of interest are recovered with high yield
- The columns are pre-filled with size exclusion matrix
- Maximum sample volume to be loaded onto the column: 20 μL
- Columns with dry gel matrix can be stored at room temperature for 12 months
- Columns with hydrated gel matrix can be stored at 4°C for 14 days
Specifications
10 Preps | |
5 min/prep (without hydration of matrix) | |
Dye terminator removal |