missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoSpin™ 96 Gel and PCR Clean-Up Kit

NucleoSpin™ 96 Gel and PCR Clean-Up Kit provides time saving medium to high throughput PCR clean up.

Brand:  Macherey-Nagel Bioanalysis™ 740658.24


 View more versions of this product

Product Code. 12745214

  • 3040.00€ / 2304 reactions
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The NucleoSpin™ 96 PCR Clean-up kit allows direct clean-up of PCR reaction mixtures. Within the procedure the addition of chaotropic salt (Buffer NT) allows a reversible adsorption of the PCR products to the silica membrane of the NucleoSpin™ 96 PCR Clean-up Binding Plates. High purity of the PCR-products is achieved by complete removal of primers, primer-dimers, salts, nucleotides, and proteins (e.g., polymerases, BSA) in subsequent washing steps using Buffer NT3. Highly pure PCR products are finally eluted with Elution Buffer NE (5 mM Tris/HCl, pH 8.5) or water (pH 8.5), and can be used directly for further applications.

  • Silica membrane technology
  • Processing: manual/automated, vacuum/centrifugation
  • Sample material: < 10 0μL PCR reaction mixture
  • Elution volume: 75 to 150 μL
  • Binding capacity: 15 μg
  • Preparation time: 45 min/plate
  • Fragment size: 65 bp to 10 kb
Specifications

Specifications

18°C to 25°C
PCR clean up
< 100 μL
96-well Plate
2304 Preps
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.