Learn More
Macherey-Nagel™ NucleoSpin™ miRNA Plasma Kit
Brand: Macherey-Nagel™ 740981.10
Specifications
Mini Spin Column | |
NucleoSpin™ miRNA Columns, Collection Tubes (2 mL), buffers, RNase-free rDNase | |
NucleoSpin™ miRNA Plasma Kit | |
<1000 nt / bp |
Isolation of small RNA from plasma or serum for cancer research | |
10 Reactions | |
Yes | |
40 min./10 Reaction and 70 min./10 Reaction |
Safety and Handling
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1