Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Brand: Thermo Scientific™ 254437
352.93 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
- Radiation sterilized—no need for flame sterilization
- Hydrophilic treatment during manufacturing process
- Liquid adherence properties that prevent accidental discharge of droplets
- Loops available in 10 and 1 ÎĽL sizes
- Meet semi-quantitative standard for handling samples
- Calibration documents available for any batch upon request
Specifications
Single-use | |
Polystyrene | |
Blue | |
Carton of 600 |
10 ÎĽL | |
Sterile | |
Inoculating Loop (Blue) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
184037 | Certificate of Irradiation | 09/01/2025 | 254437 |
184037 | Certificate of Compliance | 03/01/2025 | 254437 |
174893 | Certificate of Compliance | 03/12/2024 | 254437 |
170762 | Certificate of Compliance | 03/12/2024 | 254437 |
182714 | Certificate of Compliance | 03/12/2024 | 254437 |