Learn More
Thermo Scientific™ Nunc™ High Density Cell Factory™ System
BIOPHARMA
Designed to increase cell culture yield and improve productivity in the same footprint as a standard Thermo Scientific™ Nunc™ Cell Factory™ system.
Brand:  Thermo Scientific™ 169160
356.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16659752
  • 438.00€ / Pack of 4
Only null left
In Stock
Description
Thermo Scientific™ Nunc™ High Density Cell Factory™ System offers 30% more surface area and yield* than the standard Cell Factory system or similar multi-tray systems for adherent cell culture. Its economic benefits are immediate with impacts across many functions of an organization, as it enables end users to optimize their cell culture manufacturing footprint, reduce material usage and labor-consumption as well as to increase their cell culture yield.
  • Increase your Yield - 30% MORE surface area and yield* all within the standard Nunc Cell Factory system footprint
  • Enhance your Productivity - Increase your labor and handling efficiencies by achieving more output in a single process run
  • Improve your Process Economics - Increase your manufacturing capacity without capital investment
  • Seize Environmental Opportunities - Consume less and reduce your decontamination and waste disposal costs
*The increase in yield may vary depending on the type of cells cultured.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nunc High Density Cell Factory System
2.1 in.
4/Cs.
Less waste, renewable material
1896 cm²
Polystyrene
54 mm
1 Ea.
Vented
3
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number184591 Certificate TypeCertificate of Compliance Date09/05/2025 Catalog Number169160
Lot Number184184 Certificate TypeCertificate of Compliance Date11/01/2025 Catalog Number169160
Lot Number178832 Certificate TypeCertificate of Compliance Date06/12/2024 Catalog Number169160
Lot Number184591 Certificate TypeCertificate of Irradiation Date17/03/2025 Catalog Number169160
Lot Number184184 Certificate TypeCertificate of Irradiation Date11/01/2025 Catalog Number169160
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ High Density Cell Factory™ System > Layers: 3, Area: 1896 cm², No. Per Case: 4/Cs.