Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes
GREENER_CHOICE
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes.
Brand:  Thermo Scientific™ 4021
1095.80 EUR valid until 2025-12-31
Use promo code "24345" to get your promotional price.
Product Code. 10099092
  • 1295.00€ / Pack of 500
Estimated Shipment: 14-05-2025
Description
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes.
  • Available with a molded grid
  • Performs well in automatic dispensers
  • Contact dish useful for sampling in hospital environments as well as in food science and the pharmaceutical industry
  • Compatible with automated systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vented, without stack ring
79 sq. cm
Petri Dish
Yes
Flat
Square
1
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1336741 Certificate TypeCertificate of Compliance Date09/12/2024 Catalog Number4021
Lot Number1303884 Certificate TypeCertificate of Compliance Date09/12/2024 Catalog Number4021
Lot Number1361106 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4021
Lot Number1305898 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4021
Lot Number1396407 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4021
Product Title
Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes > Vented, without stack ring