Learn More
Masterflex™ Straight Fitting, Barb x NPT(M), Nylon
Cole-Parmer™ Straight Fitting, Barb x NPT(M), Nylon is easy to use and provides secure tubing connections.
Brand: Masterflex™ 30621-32
This item is not returnable.
View return policy
Description
- Nylon construction
- Easy to use
- Sterilizable via ETO
Specifications
Male Pipe Adapter, Nylon, 1/8 in. x 1/4 in. | |
Nylon | |
10 Pack | |
Ethylene Oxide |
Not Autoclavable | |
1/4 in. NPT(M) thread x 1/8 in. ID | |
Fits tubing accepting 1/4 in. NPT connection, 1/8 in. ID tubing | |
1/4 in. NPT(M) |