Learn More
Bollé Safety Cobra Platinum™ Clear Safety Glasses
Click to view available options
Clear
Description
- The option of swapping the temples for an adjustable stra and foam seal
- Upper protection
- Panoramic vision
- Non-slip bridge
- Pivoting temples
Specifications
Specifications
Certifications/Compliance | European standard |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Glasses |
Includes | Option of swapping temples for an adjustable strap and foam seal |
No. per Pack | 1 Ea. |
Frame Material | Nylon, Polycarbonate, TPR |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Videos
-
Bolle Safety Platinum efficiency demonstration
-
Bolle Safety Cobra
-
Bolle Safety Platinum coating
Documents
Product Information
Specification Sheets
COBRA
File size: 1.2 MB
Promotions
-
Multiple Manufacturers
Safety Monthly Promotion
Discover exclusive monthly deals on essential safety products to enhance your lab's protection and compliance.Promo Code: 24260Expires: 2025-05-31
New Offer Save Up to 76% -
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
Bollé Safety Cobra Platinum™ Clear Safety Glasses >