Learn More
Motic™ Objective Lens for SMZ160
The Objective Lens is compatible with the SMZ-160 microscope.
Brand: Motic™ 1101001708271
This item is not returnable.
View return policy
Description
- Objective Lens compatible with SMZ160.
- Not compatible with standard stands.
- Available in sizes 0.35x with WD = 287 mm, 0.5x with WD = 165 mm, 0.75x with WD = 120 mm, 1.5x with WD = 47 mm and 2.0 with WD = 30 mm.
Specifications
SMZ160 Objective | |
Reduce zoom range of SMZ-160 | |
0.75X | |
120 mm |
120 mm | |
SMZ-160 | |
5 years |