Learn More
OHAUS™ Ethernet Interface Kit for Explorer™ Precision High Capacity Balance
For Explorer Precision High Capacity Balance.
Brand:  OHAUS™ 83021082
148.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11820133
  • 156.00€ / Each
Description
For use with Explorer Precision High Capacity Balance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Explorer Precision High Capacity Balance
Ethernet Interface Kit
Videos
  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Ohaus Explorer balances
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    OHAUS™ Ethernet Interface Kit for Explorer™ Precision High Capacity Balance > Ethernet Interface for Explorer Precision