Learn More
OHAUS™ Security Lock-down Device for Explorer™ Pro Toploading and Analytical Balances
For use with Explorer Pro Toploading and Analytical Balances.
Brand:  OHAUS™ 80850043
217.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10496791
  • 226.00€ / Each
Description
  • For use with Explorer Pro Toploading and Analytical Balances
  • Security laptop lock with keys
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Explorer Pro Toploading and Analytical Balance
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Security Laptop Lock
Keys
Anti-Theft Devices
Videos
  • Ohaus - Explorer balances
    Ohaus - Explorer balances
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Ohaus Explorer balances
Product Title
OHAUS™ Security Lock-down Device for Explorer™ Pro Toploading and Analytical Balances > Security Lock-down Device