missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Olopatadine hydrochloride

Product Code. 17251226
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140462-76-6
Molecular Formula C21H24ClNO3

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: soluble.
Melting Point 248.0°C
Formula Weight 373.88
InChI Key HVRLZEKDTUEKQH-NOILCQHBSA-N
SMILES [H+].[Cl-].CN(C)CC\C=C1\C2=CC=CC=C2COC2=CC=C(CC(O)=O)C=C12
IUPAC Name hydrogen 2-[(2Z)-2-[3-(dimethylamino)propylidene]-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-5-yl]acetic acid chloride
Molecular Weight (g/mol) 373.88
Chemical Name or Material Olopatadine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'