Learn More
B Braun™ Ominifix™ Solo Luer Lock Cone
Brand:  B Braun™ 4617509F
Product Code. 12702687
  • 163.00€ / Pack of 100
Specifications
EN ISO 7886-1
Centric, Luer Lock
1/Ea.
Sterile
Polypropylene
Single-use
50 mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
B Braun™ Ominifix™ Solo Luer Lock Cone > Capacity: 50mL