Learn More
Honeywell Safety Products™ OP-TEMA Safety Glasses
Click to view available options
Lens Coating: Anti-Scratch
Description
- Affordably priced protection for maximum value.
- Lightweight full-view lens.
- Built-in molded browbar.
- Soft, adjustable nosepiece.
- Adjustable temple length and lens angle.
- Abrasion resistant polycarbonate lens.
- Adjustable, breakaway Flexicord retainer.
- Added security and convenience in the active work environment.
- Unique 9 base curved lens design.
- Excellent optical qualities and complete protection.
Specifications
Specifications
Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC0723, EC Attestation Number EC0723 |
Lens Coating | Impact-resistant, Scratch-resistant, UV-treated |
Product Type | Safety Glasses |
For Use With (Application) | Administration |
Frame Color | Burgundy |
Includes | Adjustable, breakaway Flexicord retainer |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Nylon |
UV Protection | UV400 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Product Content Correction
Product Title
Honeywell Safety Products™ OP-TEMA Safety Glasses >