Learn More
BRAND™ Optical Glass Cavity Slides
With rectangular ground edges.
Brand: BRAND™ 475505
143.75 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- White (clear) soda-lime glass, hydrolytic class 3
- Single concavity
- Provided in packs of 50
Specifications
50 | |
Soda Lime Glass | |
Optical Glass Slide | |
Clear White |
Ground | |
1.2 to 1.5 mm | |
1 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%