Learn More
Thermo Scientific™ Orion™ 9103BNWP Combination pH Electrode
Conduct applications with the accuracy and broad temperature range of this pH electrode, which includes widely used silver/silver chloride references.
Brand:  Thermo Scientific™ 9103SC
 View more versions of this product
Product Code. 10738591
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
Conduct applications with the accuracy and broad temperature range of the Thermo Scientific™ Orion™ 9103BNWP Combination pH Electrode, which includes widely used silver/silver chloride references.
Includes:
  • 60ml bottle of Orion filling solution, 900011
TRUSTED_SUSTAINABILITY
Specifications
Specifications
With screw cap connector
Ceramic
0 to 14
Combination pH Electrode
Ag/AgCl
32°F to 194°F
Screw Cap
Glass Body
0°C to 90°C
4M KCl with Ag/AgCl
0.02
16 mm