Learn More
Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe
Reduce probe servicing time and eliminate assembly problems using the Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe.
60.10€ - 117.00€
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
10654603
|
Thermo Scientific™
080515 |
Orion DO Probes 083005MD, 083010MD, 083025MD, 083060MD, 086030MD, 083005A, 083010A, 083005D, 086020A, 081010F | 1 cap |
60.10€
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||||
11767757
|
Thermo Scientific™
081003 |
Orion DO Probes 080510MD, 081010MD, 081010 | Three Caps | N/A | |||||
Description
Reduce probe servicing time and eliminate assembly problems using Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe. Convenient screw-on membrane caps help ensure optimal probe performance.
Warranty: Out of boxSpecifications
Dissolved Oxygen Probe Accessories | |
Polarographic DO |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.