Learn More
Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case with Orion™ Star A portable meters to protect, transport and store equipment in the field
Brand:  Thermo Scientific™ STARA-CS
Product Code. 11651489
  • 144.72€ / Each
In Stock
Description
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case with the Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series) to protect, transport and store your equipment in the field. Field case holds Star A series portable meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case plus the internal separating door can be removed for cable management.
Hard-sided field case includes cable management system for electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orion Star A series hard-sided field case
Meter Accessories
Videos
  • Thermo Scientific - Orion meters - Taking water analysis to the next level
    Antonia Finlayson introduces Thermo Scientific Orion Star A and VERSA STAR meters for water analysis spanning pH, ISE, conductivity and dissolved oxygen measurements.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific Orion meters Taking water analysis to the next level
    • Thermo Scientific - Orion meters - Taking water analysis to the next level
      Antonia Finlayson introduces Thermo Scientific Orion Star A and VERSA STAR meters for water analysis spanning pH, ISE, conductivity and dissolved oxygen measurements.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Thermo Scientific Orion meters Taking water analysis to the next level
      Product Title
      Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case > Hardside Carrying Case