Learn More
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts
Choose these replacement parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
Brand: Thermo Scientific™ A3-1-EG
This item is not returnable.
View return policy
Description
Choose Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
The gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run utilizing a wall comb. Built in buffer exchange ports allows buffer recirculation on extended runs, if needed. The large number of comb slots gives you sample and resolving distance flexibility.Specifications
Replacement end gates (1 pair) | |
Owl™ A3-1 and A6 Wide Gel System | |
Owl | |
End Gates |