Learn More
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts
Product Code. 12444537
Click to view available options
Type:
End Gates
Gasket
Gel Tray
Unit Size:
2 sets
Each
Pack of 2
Product Code. 12444537
Brand: Thermo Scientific™ A3GK1
Only null left
Add to basket
Add to basket
Choose these replacement parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
Choose Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
The gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run utilizing a wall comb. Built in buffer exchange ports allows buffer recirculation on extended runs, if needed. The large number of comb slots gives you sample and resolving distance flexibility.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ A3-1 and A6 Wide Gel System
Product Line Owl
Type Gasket
Quantity 1 Pair