Learn More
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts
Click to view available options
Type:
End Gates
Gasket
Gel Tray
Unit Size:
2 sets
Each
Pack of 2
Description
Choose Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
The gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run utilizing a wall comb. Built in buffer exchange ports allows buffer recirculation on extended runs, if needed. The large number of comb slots gives you sample and resolving distance flexibility.
Specifications
Specifications
| For Use With (Equipment) | Owl™ A3-1 and A6 Wide Gel System |
| Product Line | Owl |
| Type | Gasket |
| Quantity | 1 Pair |