Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
25.55€ - 207.00€
Specifications
For Use With (Equipment) | Owl™ D2 Wide Gel Electrophoresis System |
---|---|
Product Line | Owl |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12414607
|
Thermo Scientific™
D2-CST |
External caster (no trays/combs) |
116.00€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
12424607
|
Thermo Scientific™
D2-GK |
Replacement Gaskets (1 pair) |
25.55€
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
12464607
|
Thermo Scientific™
D2-UVT |
EasyCast™ gasketed UVT gel tray |
207.00€
Each |
In Stock
Log in to see stock. |
|||||
Description
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
Specifications
Owl™ D2 Wide Gel Electrophoresis System | |
Owl |