Learn More
Oxalic Acid Solution, 0.05M, Chem Lab™ Product Code.: 10730195
Oxalic Acid Solution, 0.05M, Chem Lab™
Product Code. 10730195
1 L
Click to view available options
Quantity:
1 L
Product Code. 10730195
Brand: Chem Lab CL05.1502.1000
Specifications
Density 1,00 g/mL
Quantity 1 L
Product Content Correction
Product Title
Oxalic Acid Solution, 0.05M, Chem Lab™ >