Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sealing Clip
Use with AnaeroGen Compact No. AN0010C; Compact Plastic Pouch, No. AG0020C; Compact Sachet, No. AN0020D; Thermo Scientific™ CampyGen™ Compact Sachet, No. CN0020C.
Brand: Thermo Scientific™ AN0005C
27.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sealing Clip is for use with AnaeroGen Compact Part No. AN0010C; Compact Plastic Pouch, Part No. AG0020C; AnaeroGen Compact Sachet, Part No. AN0020D; Thermo Scientific™ CampyGen™ Compact Sachet, Part No. CN0020C.
Specifications
Compact Sealing Clip | |
Thermo Scientific Oxoid | |
5/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6269628 | Certificate of Analysis | 15/05/2025 | AN0005C |
6259113 | Certificate of Analysis | 24/04/2025 | AN0005C |
6257137 | Certificate of Analysis | 13/04/2025 | AN0005C |
6251119 | Certificate of Analysis | 22/03/2025 | AN0005C |
6226841 | Certificate of Analysis | 01/02/2025 | AN0005C |