Learn More
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods.
Brand:  Thermo Scientific™ HP0053A
41.38 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10609174
  • 42.65€ / Pack of 6
Estimated Shipment: 02-05-2025
Description
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For use only with Oxoid Antimicrobial Susceptibility Discs
Oxoid Antimicrobial Susceptibility Individual Disc Dispenser
Individual Disc Dispenser
6 Ejectors/Pk.
SDS
Documents Certificates
Product Title
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser > 6/Pk