Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide
Oxoid IMAGEN Respiratory Syncytial Virus Positive Control is a slide with acetone-fixed human epithelial cells infected with RSV.
Brand: Thermo Scientific™ S610830-2
173.94 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The IMAGEN™ Control Slides are for use as a control to the following tests: IMAGEN Adenovirus (K610011-2) IMAGEN Respiratory Syncyti al Virus (RSV) (K610211-2) IMAGEN Parainfluenza Group (K610311-2) IMAGEN Parainfluenza Typing 1, 2 and 3 (K610411-2) IMAGEN Influenza A and B Virus (K610511-2) IMAGEN Respiratory Screen (K612011-2) IMAGEN hMPV (K612511-2) IMAGEN Herpes Simplex Virus (HSV) (K610611-2) IMAGEN Chlamydia kit (K610111-2)
Sealed in foil pouch filled with nitrogen.
Specifications
IMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide | |
Control | |
1 Slide/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3568265 | Certificate of Analysis | 12/09/2023 | S610830-2 |
3403194 | Certificate of Analysis | 06/08/2022 | S610830-2 |
3398341 | Certificate of Analysis | 17/12/2021 | S610830-2 |
3374078 | Certificate of Analysis | 10/11/2021 | S610830-2 |
3361790 | Certificate of Analysis | 09/10/2021 | S610830-2 |