Learn More
Thermo Scientific™ Oxoid™ X + V Factor Disc
Oxoid X + V Factor Disc is used for identification of Haemophilus spp.
Brand: Thermo Scientific™ DD0005T
34.72 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Oxoid X + V Factor Disc is used for identification of Haemophilus spp.
50 Discs/Cartridge
Specifications
-10°C to -20°C | |
Haemophilus species | |
1 Cart./Pk. |
X + V Factor Disk | |
V Factor Disc |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6199998 | Certificate of Analysis | 22/01/2025 | DD0005T |
6145404 | Certificate of Analysis | 09/10/2024 | DD0005T |
6002061 | Certificate of Analysis | 17/05/2024 | DD0005T |
3698330 | Certificate of Analysis | 06/01/2024 | DD0005T |
3690517 | Certificate of Analysis | 06/01/2024 | DD0005T |