missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene dibromide, 97%, Thermo Scientific Chemicals

Product Code. 11461668
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in water (react), and toluene.

Notes
Incompatible with alcohols, bases, amines, oxidizing agents. Avoid moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-24-5
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000182
InChI Key RBZMSGOBSOCYHR-UHFFFAOYSA-N
Synonym 1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
PubChem CID 69335
IUPAC Name 1,4-bis(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)CBr

Specifications

Melting Point 144°C to 148°C
Density 2.012
Boiling Point 245°C
Quantity 10 g
UN Number UN3261
Beilstein 1100019
Solubility Information Soluble in water (react),and toluene.
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material p-Xylylene dibromide
compliance-icons
Product Identifier
  • p-Xylylene dibromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : p-Xylylene dibromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.