Learn More
Abnova™ p21/CEN6p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Marque:  Abnova™ FG0062.100uL
 Afficher plus de versions de ce produit
Code produit. 16093085
  • 1515.00€ / 100µL
Estimated Shipment: 18-12-2024
to see stock.
Découvrez plus d'offres spéciales
Ajouter au panier
Includes: DAPI Counterstain (1500ng/mL) 125μL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Spécification
Spécification
Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
6p21.2, 6p12.1
p21/CEN6p FISH Probe
FITC, Texas Red
DAPI Counterstain (1500 ng/mL ) 250 uL
p21, CEN6p
100 μL
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats
Offres spéciales
Offres spéciales