missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p40/deltaNp63 Rabbit anti-Human, Clone: P40/7302R, Novus Biologicals™
Rabbit Monoclonal Antibody
285.00€ - 524.00€
Specifications
Antigen | p40/deltaNp63 |
---|---|
Clone | P40/7302R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18375874
|
Bio-Techne
NBP3-13806-20UG |
20 μg |
285.00€
20µg |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18392536
|
Bio-Techne
NBP3-13806-100UG |
100 μg |
524.00€
100µg |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
p40/deltaNp63 Monoclonal antibody specifically detects p40/deltaNp63 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
p40/deltaNp63 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9H3D4 | |
8626 | |
IgG κ | |
Protein A purified |
P40/7302R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
AIS, amplified in squamous cell carcinoma, B(p51A), B(p51B), Chronic ulcerative stomatitis protein, EEC3, Keratinocyte transcription factor KET, KET, LMS, OFC8, p53CP, SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein p53-competing protein | |
Recombinant fragment (around aa1-200) of human p40/deltaNp63 protein (exact sequence is proprietary) (Uniprot: Q9H3D4 ) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title