Learn More
Dostmann Electronic™ P755 Model 2-channel Multi-function Handheld Instrument
Brand: Dostmann Electronic™ 5000-0755
This item is not returnable.
View return policy
Specifications
Areas of application: For taking measurements for quality assurance under ISO 9000, As a reference instrument for checking production | |
0 to 100% | |
P755 | |
2-channel Multi-function Handheld Instrument | |
-200°C | |
±0.03/±0.05°C or 0.05% Pt100 | |
93 mm |
44 mm | |
200 mm | |
300 g | |
850°C | |
-200°C to +850°C | |
2-line LCD |