Learn More
Palladium(II) acetate, trimer, 99.98% (metals basis), Pd 47% min Product Code.: 11374738
Palladium(II) acetate, trimer, 99.98% (metals basis), Pd 47% min
Product Code. 11374738
5 g
Click to view available options
Quantity:
1 g
5 g
Product Code. 11374738
Brand: Thermo Scientific Alfa Aesar 043086.06
Request Bulk or Custom Format
CAS: 3375-31-3 | C4H6O4Pd | 224.51 g/mol
Palladium(II) acetate is a catalyst of choice for a wide variety of reactions such as vinylation, Wacker process, Buchwald-Hartwig amination, carbonylation, oxidation, rearrangement of dienes (e.g., Cope rearrangement), C-C bond formation, reductive amination, etc. Precursor to Pd(0), other Pd(II) compounds of catalytic significance, and Pd nanowires.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3375-31-3
Molecular Formula C4H6O4Pd
Molecular Weight (g/mol) 224.51
MDL Number MFCD00012453
InChI Key YJVFFLUZDVXJQI-UHFFFAOYSA-L
Synonym palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
PubChem CID 167845
SMILES [Pd++].CC([O-])=O.CC([O-])=O
Specifications
Melting Point ∼205°C (decomposition)
Color Red to Brown
Physical Form Powder
Quantity 5 g
Linear Formula [Pd(C2H3O2)2]3
Merck Index 14,6991
Solubility Information Soluble as monomer in glacial acetic acid or as trimer in benzene.
IUPAC Name palladium(2+);diacetate
Formula Weight 673.46
Percent Purity 99.98%
Concentration or Composition (by Analyte or Components) Pd 47% min
Assay (metals basis)
Chemical Name or Material Palladium (II) acetate, Trimer
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP18L056 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number043086.06,043086.03
Lot NumberZ27K063 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number043086.06,043086.03
Lot NumberP22K059 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number043086.06,043086.03
Lot NumberZ04J002 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number043086.06,043086.03
Lot NumberN24G036 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number043086.06,043086.03
Product Title
Palladium(II) acetate, trimer, 99.98% (metals basis), Pd 47% min >