Learn More
Checkers Park-It™, Black/Yellow
The rubber Park-It™ Parking Curb is used to effectively guide cars when pulling into a parking place. It is profiled at 10 cm high, safeguarding potential damage to today’s low, front-end car designs. They are more resistant than concrete or plastic curbs.
Brand:  Checkers GNRS1410YB
 View more versions of this product
Product Code. 17722318
  • 57.00€ / Each
Estimated Shipment: 11-06-2025
Description
  • The Park-It™ parking curb is constructed of 100% recycled rubber and is used to effectively guide cars when pulling into a parking stall
  • It is profiled at 10 cm high, safeguarding potential damage to today's low, front-end car designs
  • Unlike concrete or plastic, which necessitate frequent replacements, the Park-It™'s recycled rubber construction makes it safer and more durable
  • In-moulded highway reflective tape (available in yellow or white) ensures visibility and pedestrian safety
  • The Park It™ comes with pre-moulded holes and can be shipped with rebar spikes or lag bolts for installation
  • They are lightweight (16 kg for an 18 cm Park-It™) and flexible; conforming to the contour of uneven road surfaces
  • Park-It™ parking curbs are available in 90 cm, 120 cm and 180 cm lengths and are covered by a 3-year warranty; 1 year on the tape (under normal use)
Specifications
Specifications
47 in.
15 cm
Recycled Rubber
120 cm
Black, Yellow
Parking Curb
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Checkers Park-It™, Black/Yellow > Parking Curb; Length: 120 cm