Learn More
Tocris Bioscience™ Paroxetine maleate
Highly potent and selective 5-HT uptake inhibitor
Brand:  Tocris Bioscience™ 2141/50
590.05 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18798566
  • 746.00€ / 50mg
Estimated Shipment: 04-12-2024
Description
Paroxetine maleate is a highly potent and selective 5-HT uptake inhibitor that binds with high affinity to the serotonin transporter (Ki = 0.05 nM). Ki values are 1.1, 350 and 1100 nM for inhibition of [3H]-5-HT, [3H]-l-NA and [3H]-DA uptake respectively. Displays minimal affinity for α1-, α2- or β-adrenoceptors, 5-HT2A, 5-HT1A, D2 or H1 receptors at concentrations below 1000 nM, however displays weak affinity for muscarinic ACh receptors (Ki = 42 nM). Antidepressant and anxiolytic in vivo.
Specifications
Specifications
>99%
Paroxetine maleate
Paroxetine maleate
50 mg
445.44