Learn More
Emma Safety Footwear Patrick Safety Shoes
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
Brand: Emma Safety Footwear MM73084642
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
42 |
Laces | |
Patrick | |
Leather | |
Steel |