Learn More
Emma Safety Footwear Paul XD Safety Shoes
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
Brand:  Emma Safety Footwear MM70186638
 View more versions of this product
Product Code. 15948861
  • 128.00€ / 1 pair
Estimated Shipment: 29-09-2025
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
Paul XD
Leather
38
Product Title
Emma Safety Footwear Paul XD Safety Shoes > Size: 38