missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ PBS 10X
PBS 10X, Rnase, Dnase, Protease free, Filtered through 0,2ÎĽm PTFE.
Brand: MP Biomedicals™ 11PBS10X03
This item is not returnable.
View return policy
Description
- PBS 10 X buffer is ideal for maintaining a constant pH, and since it is isotonic and non-toxic to cells.
- it can be used in a wide variety of biological applications.
- PBS 10X is frequently used as a wash and dilution buffer.
- It is also used to take a reference spectrum when measuring protein adsorption.

Specifications
7.4 L | |
Phosphate Buffered Saline (PBS) | |
Buffer, Mammalian Cell Culture |
Store at Room temperature (15-30°C). | |
5 x 1 L | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ PBS 10X > 5 x 1L
Spot an opportunity for improvement?Share a Content Correction