Learn More
Eppendorf™ PCR Rotor
Available with a wide range of adapters to add versatility to PCR applications
Brand: Eppendorf™ 5452727007
308.40 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with PCR Applications
Specifications
PCR Rotor | |
2 x PCR Strips | |
MiniSpin™/MiniSpin™ plus Centrifuges |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
89Hours
1Minutes
39Seconds
55New Offer Save Up to 85%
Product Content Correction
Product Title
Eppendorf™ PCR Rotor > 2 x PCR strips