Learn More
Tocris Bioscience™ PD 90780
Inhibits NGF binding to p75NTR
Brand: Tocris Bioscience™ 5101/10
This item is not returnable.
View return policy
Description
PD 90780 is a a nonpeptide inhibitor of NGF binding to p75NTR (IC50 ∽ 1 μM in CHO cells); binds NGF, not p75NTR. Affinity maybe altered by the presence of TrkA receptors. Neuroprotective.
Specifications
>98% | |
PD 90780 | |
PD 90780 |
10 mg | |
362.34 |
Product Content Correction
Product Title
Tocris Bioscience™ PD 90780 > 10mg