Learn More
BRAND™ LDPE Grip Stoppers
Manufactured from Low-density polyethylene (LDPE) BRAND™ LDPE Grip Stoppers are used with sample and sedimentation tubes. The dual-ringed stopper prevents sample leakage. Ridged edge provides easy gripping and handling.
Brand:  BRAND™ 114720
28.79 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 10058670
  • 37.65€ / Pack of 1000
In Stock
Description
  • Manufactured from Low-density polyethylene (LDPE)
  • For use with sample and sedimentation tubes.
  • Dual-ringed stopper prevents sample leakage.
  • Ridged edge provides easy gripping and handling.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
16mm
BRAND™ Sample Tube 114715
16 Dmm
Grip Stopper
Sample and sedimentation tubes
1000/Ea.
LDPE
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
BRAND™ LDPE Grip Stoppers > Size: 16mm