missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ PE-LD Safety Wash Bottle
Click to view available options
Capacity (Metric):
500 mL
Description
- PE-LD
- Yellow translucent
- With screwed cap PP
- Imprinted with German and English
- Name and chemical formula of intended contents, together with symbols indicating type of hazard acc. to Ordinance on Hazardous Substances
Specifications
Specifications
Capacity (Metric) | 500 mL |
Color | Transparent Yellow |
Description | Safety Wash Bottle with content indication |
Material | LDPE |
Quantity | 12/Pack |
Transparency | Yes |
Product Type | Safety Wash Bottle |
Chemical Label | Methanol |
Closure Type | Screw Cap |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'