Learn More
Brand™ Pipette Rinser
Ideal quick and safe solution for washing pipettes up to 460mm long.
Brand:  Brand™ 29120
294.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10353531
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Specifications
734 mm
170mm
PE-HDf
Automatic Pipette Washer
Product Title
Brand™ Pipette Rinser > DiameterMetric: 170mm