Learn More
Cytiva PEEK Male Connector
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
Brand: Cytiva 18-1112-49
This item is not returnable.
View return policy
Description
- Chromatography accessory
- Complete Packsize 2
- Select from a variety of connections sizes and internal diamters
- For use with chromatography tubing
- Connectors made of Polyetheretherketone (PEEK)
Specifications
Male Connector | |
3/16 in. Male | |
PEEK | |
3/16 in. O.D. Tubing |