Learn More
Thermo Scientific Chemicals Pepsin, for biochemistry, powder
Pepsin from porcine gastric mucosa, CAS # 9001-75-6, is a peptidase that is commonly used in enzymatic digestion of proteins. | CAS: 9001-75-6
Brand: Thermo Scientific Chemicals 417075000
386.43 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Quantity | 500 g |
---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Pepsin is an endopeptidase that digests proteins by hydrolyzing peptide bonds
- It is the active form of pepsinogen, which is produced in the chief cells of the gastric mucosa
Applications
- Pepsin is commonly used to produce F(ab’)2 fragments from antibodies
- It has been used along with other enzymes to demonstrate the effects of fixation and enzymatic digestion in immunohistochemical assays, using paraffin-embedded tissue
- It can be used in hydrolyzing dry cervical samples in mice
- It has been used in the digestion of crude wheat gliadin
- It has been used to stimulate in vitro gastric digestion of cooked cod, cocoa mass, and dietary fiber
Chemical Identifiers
9001-75-6 |
Specifications
9001-75-6 | |
500 g | |
Pepsin |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0451115 | Certificate of Analysis | 17/05/2025 | 41707,417075000,417070025,417071000,417070200 |
A0467191 | Certificate of Analysis | 25/01/2025 | 417075000,417070025,417071000,417070200 |
A0451115 | Certificate of Analysis | 11/03/2024 | 417075000,417070025,417071000,417070200 |
A0400231 | Certificate of Analysis | 14/12/2022 | 417075000,417070025,417071000,417070200 |
A0443038 | Certificate of Analysis | 15/06/2022 |