missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen A4/PGA4 Antibody (03), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12534MFV450
This item is not returnable.
View return policy
Description
Pepsinogen A4/PGA4 Monoclonal antibody specifically detects Pepsinogen A4/PGA4 in Human samples. It is validated for ELISASpecifications
Pepsinogen A4/PGA4 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen A4/PGA4 (Accession#: NP_001073276.1; Met1-Ala388). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
03 | |
ELISA | |
Mouse | |
Protein A purified | |
RUO | |
643847 | |
Store at 4C in the dark. | |
IgG2a |