missing translation for 'onlineSavingsMsg'
Learn More
Percoll™, MP Biomedicals Product Code.: 11435332

Percoll™, MP Biomedicals

Product Code. 11435332 Shop All MP Biomedical Products
25 mL
Click to view available options
Quantity:
25 mL
500 mL
This item is not returnable. View return policy

Product Code. 11435332

Brand: MP Biomedicals 0219536925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 65455-52-9

Specifications

Quantity 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Percoll™, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.