Description
- Protection from splashes, flying particles
- Can be affixed to a safety helmet
- Permits use of ear muffs
- Lightweight with clear crown and chin guard for chin and neck protection
- Headgear with ratchet suspension
Specifications
Specifications
Lens Tint | Clear |
Product Type | Faceshield |
For Use With (Application) | Protection from splashes and flying particles |
Lens Material | Polycarbonate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Product Content Correction
Product Title
Honeywell™ Perforama Nova Face Shield >