missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ GC inlet liner taper for PerkinElmer instruments 4mm I.D. SGE
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 0920990
This item is not returnable.
View return policy
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with PerkinElmer instruments
Specifications
5/Pk. | |
GC Inlet Liner | |
6.2 mm |
92 mm | |
4 mm |