Learn More
Azpack™ Permanent Marker
Writes indelibly on clean dry surfaces. Azpack™ Permanent Marker can be left uncapped for several days without drying out due to dry safe technology.
Brand:  Azpack™ 11318113
Product Code. 11318113
  • 101.00€ / Pack of 10
Estimated Shipment: 11-06-2025
Description
  • DRY SAFE: The pen can be left uncapped for days without drying up (as per dry test ISO554)
  • For use on overhead film, CDs, DVDs, paper, cardboard, plastic, metal, polystyrene, glass*, porcelain, wood, leather and stone.
  • Airplane-safe: An automatic pressure equalization prevents the pen from leaking on board aircraft.
  • Permanent*, low odor ink.
*NOTE: Must be used on clean dry surfaces. Ink is not waterproof or to be used in humid conditions
Specifications
Specifications
0.8 mm
Permanent Marker
Black
10 Pk.
Documents
One moment while we fetch your results.
Product Title
Azpack™ Permanent Marker > Width: 0.8mm